Learn More
Electron Microscopy Sciences Syringe Assortment Pack
SDP
Catalog No. 50949631
Encompass_Preferred
Multiple size syringe pack used with our repetitive syringe dispenser for dispensing multiple doses of a set volume. 20 each of 0.6, 1.5, 6.0, 15, and 60 ml syringes.
Catalog No. 50-949-631 Supplier Electron Microscopy Sciences Supplier No. 72489
Multiple size syringe pack used with our repetitive syringe dispenser for dispensing multiple doses of a set volume. 20 each of 0.6, 1.5, 6.0, 15, and 60 ml syringes.