Learn More
Med Vet International Syringe Caps. Polypropylene. Non-Sterile, Fits All Luer Slip And Luer Lock Syringes. White, 10/Bag
Catalog No. 502830882
Encompass_Preferred
luer lock caps for dispensing syringes, These caps used to - for sealing syringes used in crafts, Industrials, glue sealing. These caps are compatible for luer lock syringes and luer slip syringes both. Perfect for air tight seal cap to prevent contents from drying up. For veterinary and laboratory use only, non-sterile.
Catalog No. 50-283-0882 Supplier Med Vet International Supplier No. TIPCAPWHITE10EA
luer lock caps for dispensing syringes, These caps used to - for sealing syringes used in crafts, Industrials, glue sealing. These caps are compatible for luer lock syringes and luer slip syringes both. Perfect for air tight seal cap to prevent contents from drying up. For veterinary and laboratory use only, non-sterile.