Learn More
Trajan Scientific Syringe Dispenser 10mL Gas Tight 1/EA
SDP
Catalog No. 5000494456
Encompass_Preferred
10 mL syringe with gas tight plunger and 1/4-28 UNF termination
Catalog No. 50-004-94456 Supplier Trajan Scientific Supplier No. 008987
10 mL syringe with gas tight plunger and 1/4-28 UNF termination