Learn More
Trajan Scientific Syringe Dispenser 1mL Gas Tight 1/EA
SDP
Catalog No. 5000494710
Encompass_Preferred
1 mL syringe with gas tight plunger and 1/4-28 UNF termination
Catalog No. 50-004-94710 Supplier Trajan Scientific Supplier No. 008185
1 mL syringe with gas tight plunger and 1/4-28 UNF termination