Learn More
Trajan Scientific Syringe Dispenser 2.5mL Gas Tight 1/EA
SDP
Catalog No. 5000494481
Encompass_Preferred
2 5 mL syringe with gas tight plunger and 1/4-28 UNF termination
Catalog No. 50-004-94481 Supplier Trajan Scientific Supplier No. 008687
2 5 mL syringe with gas tight plunger and 1/4-28 UNF termination