Learn More
Andwin Scientific SYRINGE DISPOSABLE 2.5ML 100/P
SDP
Catalog No. NC9805571
Encompass_Preferred
Norm-Ject Syringe, Medical-grade plastic. Polypropylene barrel. Polyethylene plunger. Inert, sterile, latex free. No rubber, silicone oil, or styrene. For sampling, mixing, and filtering. Smooth drawing.
This item is not returnable. View return policy
Catalog No. NC9805571 Supplier Andwin Scientific Supplier No. Z116858
 Restricted Item
Add to Cart
Add to Cart
Norm-Ject Syringe, Medical-grade plastic. Polypropylene barrel. Polyethylene plunger. Inert, sterile, latex free. No rubber, silicone oil, or styrene. For sampling, mixing, and filtering. Smooth drawing.