Learn More
Trajan Scientific Syringe eVol 50uL PK3 3/PK
SDP
Catalog No. 5000494188
Encompass_Preferred
50 L eVol syringe with gas tight plunger and 5 cm 0 5 mm OD bevel tipped needle PK3
Catalog No. 50-004-94188 Supplier Trajan Scientific Supplier No. 2910322
50 L eVol syringe with gas tight plunger and 5 cm 0 5 mm OD bevel tipped needle PK3