Learn More
Trajan Scientific Syringe eVol 5uL Medical 1/EA
SDP
Catalog No. 5000494038
Encompass_Preferred
5 L eVol syringe with gas tight plunger and 5 cm 0 5 mm OD bevel tipped needle
Catalog No. 50-004-94038 Supplier Trajan Scientific Supplier No. 2910020
Add to Cart
Add to Cart
5 L eVol syringe with gas tight plunger and 5 cm 0 5 mm OD bevel tipped needle