Learn More
Med Vet International Enteral / Oral Syringe Vesco 60 mL Enfit Tip Without Safety, VED-660, Each
Catalog No. 503044188
Encompass_Preferred
ISO 80369-3 compliant ENFit connector designed to reduce the risk of tubing misconnections
Catalog No. 50-304-4188 Supplier Med Vet International Supplier No. 1012175EA
ISO 80369-3 compliant ENFit connector designed to reduce the risk of tubing misconnections