Learn More
Tisch Scientific Tisch Scientific Cellulose Acetate (CA) Syringe Filter, 0.22 µm, 25 mm, Luer-Lok/Luer Slip, Nonsterile, 100 Pack, SF14487
SDP
Supplier:  Tisch Scientific SF14487
Encompass
Tisch cellulose acetate syringe filters are a hydrophilic filtration option perfect for use in aqueous protein solutions and ground water filtration applications.
Catalog No. NC1292681