Learn More
Bellco Glass, Inc. Syringe Filter, 25mm Housing Diameter, 0.45um PES membrane, Sterile, Individually wrapped, 50/cs
SDP
Supplier:  Bellco Glass, Inc. 451400423
Encompass_Preferred
Syringe Filter, 25mm Housing Diameter, 0.45um PES membrane, Sterile, Individually wrapped, 50/cs. syringe filters have pore size either 0.22μm or 0.45μm, and diameter 13mm or 25mm, with membrane materials either with PES or PVDF membranes. • For sample volumes of 2mL to 100mL• Retention volume <115µL• Maximum operating temperature: 100°C autoclave at 125°C for 15 min.• Maximum operating pressure approximately 100 psi
Catalog No. 50-233-0540