Learn More
Bellco Glass, Inc. Syringe Filter Glass Fiber
SDP
Supplier:  Bellco Glass, Inc. 229768000
Encompass_Preferred
Filters are molded in the polypropylene housings providing a reliable seal without the use of adhesives. Female luerlock inlet and male luer slip outlet. Color coded by membrane type for easy identification.Specifications GF Glass Fiber Can be used as a preliminary filtering step to avoid clogging Coarse particulate removal Prefiltration Membrane Pore Size 1.1um Membrane Diameter 30mm Case Quantity 30
Catalog No. 50-232-9712