Learn More
Bellco Glass, Inc. Syringe Filter, MCE
SDP
Supplier:  Bellco Glass, Inc. 229751000
Encompass_Preferred
Filters are molded in the polypropylene housings, providing a reliable seal without the use of adhesives. Female luer-lock inlet and male luer slip outlet.\n\nSpecifications:\n
    \n
  • Color coded by membrane type for easy identification
  • \n
  • MCE (Mixed Cellulose Ester)
  • \n
  • Filtration of aqueous solutions
  • \n
  • Effectively binds trace proteins
  • \n
  • Oil particulates and bacteria filtration
  • \n
  • Membrane Pore Size: 0.22um
  • \n
  • Membrane Diameter: 30mm
  • \n
  • Case Quantity: 30
  • \n
Catalog No. 50-232-9464