Learn More
STEMCELL Technologies Syringe Filter, Nylon, 0.22 μm, 13 mm, Size: 75 Units
SDP
Supplier:  STEMCELL Technologies 2000588
Encompass
Remove fine particles and unwanted materials with syringe filters. The nylon membrane of these filters is ideal for universal analytical procedures and chemical filtration. Each individually wrapped syringe filter is sterile non-pyrogenic and RNase/DNase-free and features a housing made with virgin polypropylene. They have a female luer lock inlet and a male luer slip outlet.
Catalog No. 50-239-0123