Learn More
Thomas Scientific Syringe Filter, Nylon, 0.22æm, 30mm Diameter
SDP
Supplier:  Thomas Scientific 1159T92
Encompass
30/cs Available in a variety of membrane types, porosities and diameters Color-coded for easy recognition Filters include luer-lock connections with polypropylene housings Filters are molded in the housings, providing a reliable seal without the use of adhesives Individual peel-to-open paper/plastic blister packs Sterile — gamma irradiated Denville Scientific Syringe Filters are used for the filtration of liquid samples that contain insoluble impurities and are made from high quality materials with very low levels of extractables. The housing material is high quality polypropylene which is chemically resistant to a wide range of solutes and solvents. Denville Scientific Syringe Filters are constructed so that the sample must pass through the membrane, not leak around it. Denville Syringe Filters are easy to use. They may be used with any luer-lock syringe and are color-coded for easy recognition. The 30mm syringe filters can filter up to 100mL of sample with a retention volume of les
Catalog No. 50-145-9628