Learn More
Azzota Corp Syringe Filter Nylon, Diameter: 33mm, Pore Size: 0.22um, 100/Pk
Supplier:  Azzota Corp AF330122
Encompass_Preferred
Syringe Filter Nylon, Diameter: 33mm, Pore Size: 0.22um, 100/pk
Catalog No. 50-161-2760