Learn More
Bellco Glass, Inc. Syringe Filter, PTFE, 0.22um
SDP
Supplier:  Bellco Glass, Inc. 229778000
Encompass_Preferred
Filters are molded in the polypropylene housings, providing a reliable seal without the use of adhesives. Female luer-lock inlet and male luer slip outlet.\n\nSpecifications:\n
    \n
  • Color coded by membrane type for easy identification
  • \n
  • PTFE (Polytetrafluoroethylene)
  • \n
  • Hydrophobic membrane has great temperature resistance
  • \n
  • Degassing/clarifying aqueous samples
  • \n
  • Strong acid solvent filtration
  • \n
  • Alkali solvent filtration
  • \n
  • Membrane Pore Size: 0.22um
  • \n
  • Membrane Diameter: 30mm
  • \n
  • Case Quantity: 100
  • \n
Catalog No. 50-233-0367