Learn More
Bellco Glass, Inc. Syringe Filter PTFE 0.45um
SDP
Supplier:  Bellco Glass, Inc. 229759000
Encompass_Preferred
Filters are molded in the polypropylene housings providing a reliable seal without the use of adhesives. Female luerlock inlet and male luer slip outlet.Specifications Color coded by membrane type for easy identification PTFE Polytetrafluoroethylene Hydrophobic membrane has great temperature resistance Degassing/clarifying aqueous samples Strong acid solvent filtration Alkali solvent filtration Membrane Pore Size 0.45um Membrane Diameter 30mm Case Quantity 30
Catalog No. 50-232-9465