Learn More
Bellco Glass, Inc. Syringe Filter PVDF
SDP
Supplier:  Bellco Glass, Inc. 229741000
Encompass_Preferred
Filters are molded in the polypropylene housings providing a reliable seal without the use of adhesives. Female luerlock inlet and male luer slip outlet. Low proteinbinding for filtration of nonaggressive aqueous and mild organic solutions.Specifications Color coded by membrane type for easy identification PVDF Polyvinylidene Fluoride Maximizes protein recovery High temperature filtration Membrane Pore Size 0.10um Membrane Diameter 30mm Case Quantity 30
Catalog No. 50-233-0041