Learn More
STEMCELL Technologies Syringe Filter, SFCA, 0.45 μm, 30 mm, Size: 30 Units
SDP
Supplier:  STEMCELL Technologies 2000587
Encompass
Remove fine particles and unwanted materials with syringe filters. The surfactant-free cellulose acetate (SFCA) membrane of these filters is ideal for the filtration of aqueous solutions such as buffers media and reagents. They contain no pre-wetting agents and exhibit low protein binding reduced extractables and low hold-up volume. Each individually wrapped syringe filter is sterile non-pyrogenic and RNase/DNase-free and features a housing made with virgin polypropylene. They have a female luer lock inlet and a male luer slip outlet.
Catalog No. 50-239-0169