Learn More
Electron Microscopy Sciences Syringe Filter SFCA, 26 mm, 45 µm pores, Acrylic Housing

Supplier: Electron Microscopy Sciences 67006
Syringe filters with high burst strength for assured performance. Manufactured to ISO 9002 and GMP standard and 100% integrity tested. These disposable filter units are ideal for tissue culture media, buffer solution, stain solution, biological fluids and most other aqueous solutions and reagents that are used daily in the EM lab. Pore size and membrane type are printed on every syringe and the filters are certified non-cytotoxic and non-pyrogenic.
Diameter: 26 mm
Pore size: 45 µm
Housing: Acrylic
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.