Learn More
Perkin Elmer US LLC for use with PerkinElmer ICP-OES and ICP-MS instruments
Supplier:  Perkin Elmer US LLC N8121035
Encompass_Preferred
Kit includes a 3 mL syringe and two luer to 1/4-28 adapters.
Catalog No. 50-210-4639