Learn More
Waters Corp Waters Syringe, 250 μL, with Shrink Wrap Plunger
Supplier:  Waters Corp WAT073109
Encompass_Preferred
This replacement 250μL syringe assembly is manufactured with precise specifications for barrel size, shape, and wall smoothness, ensuring maximum accuracy and consistency in injection volume. The syringe features a plunger seal designed for the Waters autosampler and a top seal made of unique material, providing a reliable, leak-free connection. It is utilized in Waters 717 Autosampler and in the 2695, 2695D Separations Modules.
  • 250μL volume.
  • Precise specifications for barrel, shape, and wall smoothness.
  • Ensures maximum accuracy and consistent injection volume.
  • Plunger seal designed for autosamplers.
  • Top seal of unique material for reliable, leak-free connection.
  • Compatible with 717 Autosampler, 2695, 2695 D, and 2795 Separations Modules.
  • Suitable for HPLC systems.
  • Annual replacement recommended.
Catalog No. 50-819-033