Learn More
Trajan Scientific SYRINGE GAS TIGHT 100F-LL-GT
SDP
Catalog No. 502530332
Encompass_Preferred
SGE Gastight Syringes: Fixed Luer-Lok Models, Volume: 100 uL, Diameter Outer: 0.31 in., 8 mm, Model: 100F-LL-GT, Needle Gauge: No Needle Included, Accuracy: +/-1%, Connection Type: Fixed, For Use With: Suitable for gases or liquids for maximum inertness
Catalog No. 50-253-0332 Supplier Trajan Scientific Supplier No. 005230
SGE Gastight Syringes: Fixed Luer-Lok Models, Volume: 100 uL, Diameter Outer: 0.31 in., 8 mm, Model: 100F-LL-GT, Needle Gauge: No Needle Included, Accuracy: +/-1%, Connection Type: Fixed, For Use With: Suitable for gases or liquids for maximum inertness