Learn More
Trajan Scientific SYRINGE GAS TIGHT 250F-LL-GT
SDP
Catalog No. 502530321
Encompass_Preferred
SGE Gastight Syringes: Fixed Luer-Lok Models, Volume: 250 uL, Diameter Outer: 0.31 in., 8 mm, Model: 250F-LL-GT, Accuracy: +/-1%, Connection Type: Fixed, For Use With: Suitable for gases or liquids for maximum inertness, Includes: Plunger protection, inert Kel-F male luer cone
Catalog No. 50-253-0321 Supplier Trajan Scientific Supplier No. 006230
SGE Gastight Syringes: Fixed Luer-Lok Models, Volume: 250 uL, Diameter Outer: 0.31 in., 8 mm, Model: 250F-LL-GT, Accuracy: +/-1%, Connection Type: Fixed, For Use With: Suitable for gases or liquids for maximum inertness, Includes: Plunger protection, inert Kel-F male luer cone