Learn More
Trajan Scientific SYRINGE GAS TIGHT 250R-GT
SDP
Catalog No. 502550769
Encompass_Preferred
SGE Gastight Syringes: Removable-Needle Models, Diameter Outer: 0.31 in., 8 mm, Needle Gauge: 25 G, Volume: 250 uL, Needle Point Style: Removable Bevel, Length Needle: 50 mm, 1.96 in., Accuracy: +/-1%, Connection Type: Removable, Diameter Outer Needle: 0.01 in., 0.47 mm
Catalog No. 50-255-0769 Supplier Trajan Scientific Supplier No. 006250
SGE Gastight Syringes: Removable-Needle Models, Diameter Outer: 0.31 in., 8 mm, Needle Gauge: 25 G, Volume: 250 uL, Needle Point Style: Removable Bevel, Length Needle: 50 mm, 1.96 in., Accuracy: +/-1%, Connection Type: Removable, Diameter Outer Needle: 0.01 in., 0.47 mm