Learn More
Trajan Scientific SYRINGE GAS TIGHT 500F-LL-GT
SDP
Catalog No. 502530315
Encompass_Preferred
SGE Gastight Syringes: Fixed Luer-Lok Models, Volume: 500 uL, Diameter Outer: 0.31 in., 8 mm, Model: 500F-LL-GT, Accuracy: +/-1%, Connection Type: Fixed, For Use With: Suitable for gases or liquids for maximum inertness, Includes: Plunger protection, inert Kel-F male luer cone
Catalog No. 50-253-0315 Supplier Trajan Scientific Supplier No. 007230
SGE Gastight Syringes: Fixed Luer-Lok Models, Volume: 500 uL, Diameter Outer: 0.31 in., 8 mm, Model: 500F-LL-GT, Accuracy: +/-1%, Connection Type: Fixed, For Use With: Suitable for gases or liquids for maximum inertness, Includes: Plunger protection, inert Kel-F male luer cone