Learn More
Trajan Scientific Syringe Gas tight GT 100mL 1/EA
SDP
Catalog No. 5000494526
Encompass_Preferred
100 mL syringe with removable Luer Lock valve and gas tight plunger
Catalog No. 50-004-94526 Supplier Trajan Scientific Supplier No. 009770
100 mL syringe with removable Luer Lock valve and gas tight plunger