Learn More
Trajan Scientific Syringe Gas tight GT 100uL 1/EA
SDP
Catalog No. 5000494940
Encompass_Preferred
100 L fixed needle syringe with gas tight plunger and 5 cm 0 5 mm OD bevel tipped needle calibrated
Catalog No. 50-004-94940 Supplier Trajan Scientific Supplier No. 005200CAL
100 L fixed needle syringe with gas tight plunger and 5 cm 0 5 mm OD bevel tipped needle calibrated