Learn More
Trajan Scientific Syringe Gas tight GT 100uL 1/EA
SDP
Supplier:  Trajan Scientific 005720
Encompass_Preferred
100 L fixed needle CTC syringe with gas tight plunger and 5 1 cm 0 72 mm OD (0 4 mm ID) LC tipped needle
Catalog No. 50-004-93934