Learn More
Trajan Scientific Syringe Gas tight GT 100uL 1/EA
SDP
Supplier:  Trajan Scientific 005333
Encompass_Preferred
100 L removable needle CTC Analytics and Thermo Scientific syringe with gas tight plunger and 5 cm 0 47 mm OD cone tipped needle
Catalog No. 50-004-94353