Learn More
Trajan Scientific Syringe Gas tight GT 100uL 1/EA
SDP
Supplier:  Trajan Scientific 005891
Encompass_Preferred
100 L fixed needle CTC Analytics PAL RTC (PAL3) Shimadzu AOC-6000 Thermo Scientific TriPlus RSH and Agilent PAL3 syringe with gas tight plunger and 8 5 cm 0 63 mm OD side hole needle
Catalog No. 50-004-94865