Learn More
Trajan Scientific Syringe Gas tight GT 100uL 1/EA
SDP
Supplier:  Trajan Scientific 005279
Encompass_Preferred
100 L syringe with gas tight plunger and removable needle push-pull valve with 5 cm 0 63 mm OD cone tipped needle
Catalog No. 50-004-94532