Learn More
Trajan Scientific Syringe Gas tight GT 100uL 1/EA
SDP
Supplier:  Trajan Scientific 005331
Encompass_Preferred
100 L fixed needle CTC Analytics and Thermo Scientific syringe with gas tight plunger and 5 1 cm 0 72 mm OD LC tipped needle
Catalog No. 50-004-94555