Learn More
Trajan Scientific Syringe Gas tight GT 100uL 1/EA
SDP
Supplier:  Trajan Scientific 005291
Encompass_Preferred
100 L CTC Analytics and Thermo Scientific MEPS syringe with gas tight plunger
Catalog No. 50-004-94505