Learn More
Trajan Scientific Syringe Gas tight GT 10mL 1/EA
SDP
Supplier:  Trajan Scientific 008951
Encompass_Preferred
10 mL removable needle CTC Analytics PAL RTC (PAL3) Shimadzu AOC-6000 Thermo Scientific TriPlus RSH and Agilent PAL3 syringe with gas tight plunger and 5 7 cm 1 07 mm OD LC tipped needle
Catalog No. 50-004-94558