Learn More
Trajan Scientific Syringe Gas tight GT 10uL 1/EA
SDP
Catalog No. 5000493951
Encompass_Preferred
10 L removable needle on-column syringe with gas tight plunger and 7 5 cm 0 23 mm OD removable needle
Catalog No. 50-004-93951 Supplier Trajan Scientific Supplier No. 002500
10 L removable needle on-column syringe with gas tight plunger and 7 5 cm 0 23 mm OD removable needle