Learn More
Trajan Scientific Syringe Gas tight GT 10uL 1/EA
SDP
Supplier:  Trajan Scientific 002878
Encompass_Preferred
10 L removable needle CTC RTC and Thermo RSH autosampler syringe with gas tight plunger and 8 5 cm 26 gauge 0 47 mm OD cone tipped needle
Catalog No. 50-004-94717