Learn More
Trajan Scientific Syringe Gas tight GT 10uL 1/EA
SDP
Catalog No. 5000494848
Encompass_Preferred
10 L removable needle CTC RTC and Thermo RSH autosampler syringe with gas tight plunger and 8 5 cm 23 gauge 0 63 mm OD cone tipped needle
Catalog No. 50-004-94848 Supplier Trajan Scientific Supplier No. 002874
10 L removable needle CTC RTC and Thermo RSH autosampler syringe with gas tight plunger and 8 5 cm 23 gauge 0 63 mm OD cone tipped needle