Learn More
Trajan Scientific Syringe Gas tight GT 10uL 1/EA
SDP
Catalog No. 5000494770
Encompass_Preferred
10 L removable needle CTC RTC and Thermo RSH autosampler syringe with gas tight plunger and 5 7 cm 26 gauge 0 47 mm OD cone tipped needle
Catalog No. 50-004-94770 Supplier Trajan Scientific Supplier No. 002876
10 L removable needle CTC RTC and Thermo RSH autosampler syringe with gas tight plunger and 5 7 cm 26 gauge 0 47 mm OD cone tipped needle