Learn More
Trajan Scientific Syringe Gas tight GT 10uL 1/EA
SDP
Supplier:  Trajan Scientific 002983
Encompass_Preferred
10 L fixed needle CTC Analytics syringe with gas tight plunger and 5 cm 0 72 mm OD side hole needle
Catalog No. 50-004-94480