Learn More
Trajan Scientific Syringe Gas tight GT 10uL 1/EA
SDP
Catalog No. 5000494734
Encompass_Preferred
10 L removable needle CTC Analytics PAL RTC (PAL3) Shimadzu AOC-6000 Thermo Scientific TriPlus RSH and Agilent PAL3 syringe with gas tight plunger and 5 7 cm 0 72 mm OD LC tipped needle
Catalog No. 50-004-94734 Supplier Trajan Scientific Supplier No. 002880
10 L removable needle CTC Analytics PAL RTC (PAL3) Shimadzu AOC-6000 Thermo Scientific TriPlus RSH and Agilent PAL3 syringe with gas tight plunger and 5 7 cm 0 72 mm OD LC tipped needle