Learn More
Trajan Scientific Syringe Gas tight GT 10uL 10/PK
SDP
Catalog No. 5000494757
Encompass_Preferred
10 L removable needle Agilent syringe with gas tight plunger and 4 2 cm 0 63 mm OD cone tipped needle PK10
Catalog No. 50-004-94757 Supplier Trajan Scientific Supplier No. 002820
10 L removable needle Agilent syringe with gas tight plunger and 4 2 cm 0 63 mm OD cone tipped needle PK10