Learn More
Trajan Scientific Syringe Gas tight GT 1mL 1/EA
SDP
Catalog No. 5000494856
Encompass_Preferred
1 mL removable needle CTC Analytics PAL RTC (PAL3) Shimadzu AOC-6000 Thermo Scientific TriPlus RSH and Agilent PAL3 syringe with gas tight plunger and 5 7 cm 0 72 mm OD LC tipped needle
Catalog No. 50-004-94856 Supplier Trajan Scientific Supplier No. 008151
1 mL removable needle CTC Analytics PAL RTC (PAL3) Shimadzu AOC-6000 Thermo Scientific TriPlus RSH and Agilent PAL3 syringe with gas tight plunger and 5 7 cm 0 72 mm OD LC tipped needle