Learn More
Trajan Scientific Syringe Gas tight GT 1mL 1/EA
SDP
Supplier:  Trajan Scientific 008136
Encompass_Preferred
1 mL fixed needle CTC headspace syringe with energized gas tight plunger and 5 6 cm 0 47 mm OD side hole needle
Catalog No. 50-004-94858