Learn More
Trajan Scientific Syringe Gas tight GT 1mL 1/EA
SDP
Supplier:  Trajan Scientific 008155
Encompass_Preferred
1 mL fixed needle CTC Analytics PAL RTC (PAL3) Shimadzu AOC-6000 Thermo Scientific TriPlus RSH and Agilent PAL3 headspace syringe with energized gas tight plunger and 6 5 cm 0 63 mm OD side hole needle
Catalog No. 50-004-94868