Learn More
Trajan Scientific Syringe Gas tight GT 1mL 1/EA
SDP
Supplier:  Trajan Scientific 008105CAL
Encompass_Preferred
1 mL removable needle syringe with gas tight plunger and 5 1 cm 0 72 mm OD LC tipped needle calibrated
Catalog No. 50-004-94553