Learn More
Trajan Scientific Syringe Gas tight GT 1mL 1/EA
SDP
Supplier:  Trajan Scientific 008170
Encompass_Preferred
1 mL removable needle high pressure gas tight syringe with gas tight ON/OFF valve and 7 cm 0 63 mm OD bevel tipped needle
Catalog No. 50-004-94318