Learn More
Trajan Scientific Syringe Gas tight GT 2.5mL 1/EA
SDP
Supplier:  Trajan Scientific 008505
Encompass_Preferred
2 5 mL removable needle syringe with gas tight plunger and 5 1 cm 0 72 mm OD LC tipped needle
Catalog No. 50-004-93970