Learn More
Trajan Scientific Syringe Gas tight GT 2.5mL 1/EA
SDP
Supplier:  Trajan Scientific 008636
Encompass_Preferred
2 5 mL fixed needle CTC headspace syringe with energized gas tight plunger and 5 6 cm 0 47 mm OD side hole needle
Catalog No. 50-004-94748